missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-cAMP, sodium salt, Tocris Bioscience™

Cell-permeable cAMP analog

Brand:  Tocris (Bio-Techne) 1140/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448723

  • £80.85 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell-permeable cAMP analog; activator of protein kinase A. Also available as part of the PKA Tocriset™ .
Specifications

Chemical Identifiers

76939-46-3
432.103
8-bromo-camp
sodium;(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol;hydride
C10H12BrN5NaO6P
WBHAQCJMXJAVQJ-LGVAUZIVSA-N
131954754
[H-].C1C2C(C(C(O2)N3C4=C(C(=NC=N4)N)N=C3Br)O)OP(=O)(O1)O.[Na+]

Specifications

10mg
76939-46-3
8-bromo-camp
[H-].C1C2C(C(C(O2)N3C4=C(C(=NC=N4)N)N=C3Br)O)OP(=O)(O1)O.[Na+]
432.103
430.09
8-Bromo-cAMP, sodium salt
C10H12BrN5NaO6P
WBHAQCJMXJAVQJ-LGVAUZIVSA-N
sodium;(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol;hydride
131954754
>99%
Videos
Documents