missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Azaguanine 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Color | Yellow |
Infrared Spectrum | Authentic |
Quantity | 1 g |
Merck Index | 15, 889 |
IUPAC Name | 5-amino-2H,3H,7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one |
Formula Weight | 152.12 |
Percent Purity | ≥97% |
Physical Form | Powder |
Chemical Name or Material | 8-Azaguanine, 98% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'