missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoadenine, Tocris Bioscience™

Adenine receptor agonist

Brand:  Tocris (Bio-Techne) 405050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444044

  • £382.20 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Adenine receptor agonist (Ki = 0.0341 μM in HEK293 cells expressing an adenine binding site). Displays 190-fold increased potency at the human binding site over the rat adenine receptor (rAde1R) (Ki = 6.51 μM).
Specifications

Chemical Identifiers

28128-33-8
150.145
8-aminoadenine, 1h-purine-6,8-diamine, 9h-purine-6,8-diamine
7H-purine-6,8-diamine
C5H6N6
PFUVOLUPRFCPMN-UHFFFAOYSA-N
4371597
C1=NC2=C(C(=N1)N)NC(=N2)N

Specifications

50mg
28128-33-8
8-aminoadenine, 1h-purine-6,8-diamine, 9h-purine-6,8-diamine
C1=NC2=C(C(=N1)N)NC(=N2)N
150.145
150.14
8-Aminoadenine
C5H6N6
PFUVOLUPRFCPMN-UHFFFAOYSA-N
7H-purine-6,8-diamine
4371597
>98%
Videos
Documents