missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-5-bromoisoquinoline, 97%, ACROS Organics™

Brand:  Acros Organics 431080010

Code : 29

Additional Details : CAS Number : 90721-35-0 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11946501

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

8-Amino-5-bromoisoquinoline
96.0
≥96.0%
C9H7BrN2
90721-35-0
100.0
97%
223.07

Specifications

8-Amino-5-bromoisoquinoline
96.0
Authentic
≥96.0%
186° to 190°C
C9H7BrN2
1g
90721-35-0
100.0
Crystalline Powder
97%
Off-white to brown
223.07
Safety and Handling

Safety and Handling

Product Identifier
  • 8-Amino-5-bromoisoquinoline, 97%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.

Documentation

Technical Documents

Alternative Products