missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-2H-1,4-benzoxazin-3(4H)-one, 95%, Acros Organics

Brand:  Acros Organics 445700010

Code : 29

Additional Details : CAS Number : 321126-82-3 Weight : 0.06000kg


Quantity 1g

 View more versions of this product

Product Code. 12635007

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

8-Amino-2H-1, 4-benzoxazin-3(4H)-one
95%
C8H8N2O2
164.16

Specifications

8-Amino-2H-1, 4-benzoxazin-3(4H)-one
95%
164.16
321126-82-3
C8H8N2O2
1g
Videos
SDS