missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-2-naphthol, 94%, Thermo Scientific Chemicals

Catalog Number p-3776755
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
118-46-7
C10H9NO
159.19
MFCD00004031
KVHHMYZBFBSVDI-UHFFFAOYSA-N
1-amino-7-naphthol, 8-amino-2-naphthol, 2-naphthalenol, 8-amino, 1-amino-7-hydroxynaphthalene, 7-hydroxy-1-naphthylamine, 2-naphthol, 8-amino, 8-amino-2-naphtol, 8-aminonaphth-2-ol, 1-amino-7-naphthol;, 8-amino-2-naphthaleno
8358
8-aminonaphthalen-2-ol
NC1=C2C=C(O)C=CC2=CC=C1
This item is not returnable. View return policy
118-46-7
C10H9NO
159.19
MFCD00004031
KVHHMYZBFBSVDI-UHFFFAOYSA-N
1-amino-7-naphthol, 8-amino-2-naphthol, 2-naphthalenol, 8-amino, 1-amino-7-hydroxynaphthalene, 7-hydroxy-1-naphthylamine, 2-naphthol, 8-amino, 8-amino-2-naphtol, 8-aminonaphth-2-ol, 1-amino-7-naphthol;, 8-amino-2-naphthaleno
8358
8-aminonaphthalen-2-ol
NC1=C2C=C(O)C=CC2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 92.0
CAS Max % 100.0
Melting Point 204.0°C to 207.0°C
Color Brown to Gray
Flash Point >364°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 13, 685
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 159.18
Percent Purity 94%
Physical Form Crystals or Powder
Chemical Name or Material 8-Amino-2-naphthol
Show More Show Less
compliance-icons
Product Identifier
  • 8-Amino-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'