missing translation for 'onlineSavingsMsg'
Learn More

8-(3-Chlorostyryl)caffeine, Tocris Bioscience™

Selective A2A antagonist. Also MAO-B inhibitor

Brand:  Tocris (Bio-Techne) 330650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13452754

  • £305.76 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective adenosine A2A receptor antagonist and monoamine oxidase B (MAO-B) inhibitor (Ki values are 54 and 28200 nM at rat A2A and A1 receptors respectively and Ki ∽ 100 nM at MAO-B). Potently protects against quinolinic acid -induced (Cat. No. 0225) neuronal damage and is neuroprotective in the MPTP model of Parkinson's disease.
Specifications

Chemical Identifiers

147700-11-6
661.544
1h-purine-2,6-dione,8-1e-2-3-chlorophenyl ethenyl-3,7-dihydro-1,3,7-trimethyl, acmc-20n59d
8-[2-(3-chlorophenyl)ethenyl]-1,3,7-trimethylpurine-2,6-dione
C32H30Cl2N8O4
YMUNFKYGBSLPLQ-UHFFFAOYSA-N
57348597
CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl.CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl

Specifications

50mg
147700-11-6
1h-purine-2,6-dione,8-1e-2-3-chlorophenyl ethenyl-3,7-dihydro-1,3,7-trimethyl, acmc-20n59d
CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl.CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl
661.544
330.77
8-(3-Chlorostyryl)caffeine
C32H30Cl2N8O4
YMUNFKYGBSLPLQ-UHFFFAOYSA-N
8-[2-(3-chlorophenyl)ethenyl]-1,3,7-trimethylpurine-2,6-dione
57348597
>99%
Videos
Documents