missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ 7q32.2/CEN7q FISH Probe

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

£1124.00 - £1911.00

Specifications

For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Storage Requirements Store at 4°C in the dark
Product Type DNA/RNA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16003325
View Documents
Abnova™
FG0200.200UL
200 µL
£1911.00
200µL
View alternative products
Save Up to 
»
Estimated Shipment
18-12-2020
Please to purchase this item. Need a web account? Register with us today!
16093315
View Documents
Abnova™
FG0200.100UL
100 µL
£1124.00
100µL
View alternative products
Save Up to 
»
Estimated Shipment
18-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Fluorescent In Situ Hybridization (Cell)
Store at 4°C in the dark
DNA/RNA
SDS