Learn More
Ansell™ 76-301 Series Tiger Paw™ PVC Light Weight Gloves
Catalog Number 12616991
Click to view available options
Size:
7
8
9
10
Made from polyester plated with cotton, Tiger Paw™ offers many features and benefits at an economical price. This heavier construction ensures a more robust glove for heavier applications.
  • Made from polyester plated with cotton
  • Knitted construction
  • Polyester cotton liner
  • Dotted on palm side finish
  • Knitwrist cuff
Back Material PVC
Certifications/Compliance Standard CATEGORY II EN 388 : 214X
Cuff Material PVC
Color Natural, Blue
Product Type Lightweight Gloves
Size 10
Glove Cuff Style Knit
Lining Polyester Cotton
Palm Type Dotted-palm
Quantity 12 Pairs
Series 76-301