Learn More
Applied Biosystems™ 7500 Real Time PCR Systems Spectral Calibration Kit II
This easy-to-use kit enables you to establish the pure dye spectra and multicomponent values for Cy3™, Cy5™, and Texas Red™ dyes on the 7500 Real-Time PCR System.
Brand:  Applied Biosystems™ 4351151
Code : 50
Additional Details : Weight : 0.18000kg
Product Code. 10045214
  • £499.00 / Each
Estimated Shipment
13-07-2020
For Research Use Only. All usage must comply with product instructions.
Description
Description
4349180
  • 96-well optical reaction plates preloaded with dyes (FAM™/SYBR™ Green I, VIC™/JOE™, NED™/TAMRA™/ROX™) are provided for quick and easy calibration of the 7500 Real-Time PCR System
  • Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
  • Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions
4351151
  • 96-well optical reaction plates preloaded with Cy3™, Cy5™, and Texas Red™ dyes allow quick and easy calibration of the 7500 Real-Time PCR System
  • Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously
  •  
    Spend Your Time on Research, Not Instrument Calibration:
    This kit is easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 7500 Real-Time PCR System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
    For Research Use Only. Not for use in diagnostic procedures. 
    Order Info
    Expiration date: 12 months from the date of delivery.
    Specifications
    Specifications
    Contains three Optical 96-Well Reaction Plates, preloaded and sealed containing Cy3® Texas Red® and Cy5® fluorescent dye standards.
    Store at -15 to -25°C. (This product is light sensitive.)
    Expiration date: 12 months from the date of delivery
    Cy™3, Cy™5, Texas Red®
    Spectral Calibration
    Contains three Optical 96-well Reaction Plates, preloaded and sealed containing Cy3™ Texas Red™ and Cy5™ fluorescent dye standards.
    Fluorescent
    Dry Ice
    7500 System