Learn More
Ansell™ 70-225 Series Yellow Neptune™ Heavy Weight Gloves
Catalog Number p-8004624
Click to view available options
Size:
7
8
10
100% aramid fiber construction offers a very high cut resistance to weight ratio. Repels liquids, avoiding damage to the strands and extending glove life.
  • Made from high-tech Kevlar™ Aramid fibers para-aramid short fibers
  • Neptune coating allows glove fibers to repel liquids
  • Seamless construction to avoid irritation
  • Short fibers provide extra comfort
  • Terry-loop technique enchances heat insulation
  • Thick wall offers high shock absorption
  • Ambidextrous
  • Washable and reusable
Back Material Neptune™
Certifications/Compliance Standards CATEGORY III EN 388 : 254X, EN 407, ANSI
Cuff Material Neptune™
Product Type Heavyweight Gloves
Size 10
Glove Cuff Style Knit
Liner Material Dupont™ Kevlar™ aramid fibers
Lining Dupont™ Kevlar™ aramid fibers
Quantity 144 Pairs
Resistant To Very high cut resistance