Learn More
Ansell Edmont™ Neptune™ Kevlar™ Industrial Heavy-Weight Gloves
Provide improved sensitivity and fit with increased mechanical strength.
Brand:  Ansell Edmont™ 70225070
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12646921
  • £138.13 / 12 pairs
Estimated Shipment
08-02-2018
Add to basket
Description
Description
  • Made from high-tech Kevlar para-aramid short fibers
  • Neptune coating allows glove fibers to repel liquids
  • Seamless construction to avoid irritation
  • Short fibers provide extra comfort
  • Terry-loop technique enchances heat insulation
  • Thick wall offers high shock absorption
  • Ambidextrous
  • Washable and reusable
Specifications
Specifications
Neptune®
Knitwrist
Kevlar®
7
Standards CATEGORY III EN 388 : 254X, EN 407, ANSI
Neptune®
Kevlar®
Very high cut resistance
144 Pairs
Heavy Weight Gloves
Videos