Learn More
Ansell Edmont™ 70-216 Series Yellow Neptune™ Medium Weight Gloves
Offers very high cut resistance that comes from Kevlar™ reinforcement with enhanced comfort and dexterity for your workforce. Neptune™ coating repels liquids, avoiding damage to the Kevlar strands and extending glove life.
Brand:  Ansell Edmont™ 70216070
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12600982
  • £97.60 / 12 pairs
Estimated Shipment
15-06-2018
Add to basket
Description
Description
  • Kevlar reinforcement provides high cut resistance
  • Neptune coating allows glove fibers to repel liquids
  • Reinforced thumb welt increases abrasion resistance
  • Ambidextrous
  • Washable and reusable
Specifications
Specifications
Neptune®
Knitwrist
Kevlar®
7
Standards CATEGORY III EN 388 : 144X, EN 407, ANSI
Neptune®
Kevlar®
Very high cut resistance
144 Pairs
Medium Weight Gloves
Videos