missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 7-Nitro-3-thiocyanatoindole, 97%

Brand:  Alfa Aesar™ H63988.03

Code : NEW

Additional Details : CAS Number : 885266-79-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15460989

  • £76.53 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

7-Nitro-3-thiocyanatoindole
219.22
7-Nitroindol-3-yl thiocyanate
885266-79-5
MFCD04973971

Specifications

7-Nitro-3-thiocyanatoindole
885266-79-5
97%
MFCD04973971
1g
Solid
219.22
7-Nitroindol-3-yl thiocyanate
Videos
SDS