Learn More
7-NINA, Tocris Bioscience™
Non-selective NOS inhibitor. Sodium salt of 7-nitroindazole (Cat. No. 0602)
Brand:  Tocris (Bio-Techne) 080050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13448023
  • £280.77 / 50mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Sodium salt of the non-selective NOS inhibitor 7-nitroindazole (Cat. No. 0602).
Specifications
Chemical Identifiers
161467-34-1
185.118
7-nina, 7-nitroindazole sodium salt, 7-nitroindazole monosodium salt
sodium;7-nitroindazol-2-ide
C7H4N3NaO2
DJMLKKIIBMJGIQ-UHFFFAOYSA-N
23661758
C1=CC2=C[N-]N=C2C(=C1)[N+](=O)[O-].[Na+]
Specifications
50mg
161467-34-1
7-nina, 7-nitroindazole sodium salt, 7-nitroindazole monosodium salt
C1=CC2=C[N-]N=C2C(=C1)[N+](=O)[O-].[Na+]
185.118
185.12
7-NINA
C7H4N3NaO2
DJMLKKIIBMJGIQ-UHFFFAOYSA-N
sodium;7-nitroindazol-2-ide
23661758
>98%