missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxyindole, 97%, Thermo Scientific Chemicals

Catalog Number 15460629
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3189-22-8
C9H9NO
147.177
MFCD00047203
FSOPPXYMWZOKRM-UHFFFAOYSA-N
7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi
76660
7-methoxy-1H-indole
COC1=CC=CC2=C1NC=C2
This item is not returnable. View return policy
3189-22-8
C9H9NO
147.177
MFCD00047203
FSOPPXYMWZOKRM-UHFFFAOYSA-N
7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi
76660
7-methoxy-1H-indole
COC1=CC=CC2=C1NC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.126 g/mL
Boiling Point 108°C to 110°C (0.3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.612
Quantity 1 g
Formula Weight 147.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Methoxyindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'