7-Methoxy-2-tetralone, 95%, ACROS Organics™


Overview
£136.88 valid until 31/03/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 294970250

25GR 7-Methoxy-2-tetralone, 95%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 4133-34-0 Weight: 0.07500kg


Packaging Glass bottle
Quantity 25g

Product Code. 10792315

Quantity Price
1 £ 342.2 / 25g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 7-Methoxy-2-tetralone
CAS 4133-34-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H12O2
Formula Weight 176.22
MDL Number MFCD00001730
Synonym 3, 4-Dihydro-7-methoxy-2(1H)-naphthalenone

Specification

Assay ≥94%
Physical Form Liquid After Melting
Formula Weight 176.22
Quantity 25g
Color Clear yellow to orange
CAS Min % 94.0
Infrared Spectrum Authentic
CAS 4133-34-0
Refractive Index 1.5590 to 1.5630 (20°C, 589nm)
Assay Percent Range 95%
MDL Number MFCD00001730
Packaging Glass bottle
Density 1.1300g/mL
Molecular Formula C11H12O2
CAS Max % 100.0
Chemical Name or Material 7-Methoxy-2-tetralone
Synonym 3, 4-Dihydro-7-methoxy-2(1H)-naphthalenone
Documentation

Technical documents