missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-1-tetralone, 97%, ACROS Organics™

£13.82 - £124.58

Chemical Identifiers

CAS 6836-19-7
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.21
MDL Number MFCD00001696
InChI Key GABLTKRIYDNDIN-UHFFFAOYSA-N
Synonym 7-methoxy-1-tetralone, 7-methoxyl-1-tetralone, 7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 7-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxytetralin-1-one, 3,4-dihydro-7-methoxy-1 2h-naphthalenone, 7-methoxy-2,3,4-trihydronaphthalen-1-one, 7-methoxytetralone, 7-methoxy-tetralone
PubChem CID 81276
IUPAC Name 7-methoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(CCCC2=O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10021324
SDS View Documents
Acros Organics
172430010
1g Glass bottle
£13.82
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
10547644
SDS View Documents
Acros Organics
172430050
5g Glass bottle
£33.46
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
10234734
SDS View Documents
Acros Organics
172430250
25g Glass bottle
£124.58
25g
View alternative products
Save Up to 
»
Estimated Shipment
12-11-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

6836-19-7
176.21
GABLTKRIYDNDIN-UHFFFAOYSA-N
81276
COC1=CC2=C(CCCC2=O)C=C1
C11H12O2
MFCD00001696
7-methoxy-1-tetralone, 7-methoxyl-1-tetralone, 7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 7-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxytetralin-1-one, 3,4-dihydro-7-methoxy-1 2h-naphthalenone, 7-methoxy-2,3,4-trihydronaphthalen-1-one, 7-methoxytetralone, 7-methoxy-tetralone
7-methoxy-3,4-dihydro-2H-naphthalen-1-one

Specifications

7-Methoxy-1-tetralone
Glass bottle
61°C to 63°C
6836-19-7
100.0
C11H12O2
09, II, 889
GABLTKRIYDNDIN-UHFFFAOYSA-N
7-methoxy-3,4-dihydro-2H-naphthalen-1-one
81276
Crystalline Powder
Authentic
White to Beige
1g
96.0
96% min. (GC)
MFCD00001696
7-methoxy-1-tetralone, 7-methoxyl-1-tetralone, 7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 7-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxytetralin-1-one, 3,4-dihydro-7-methoxy-1 2h-naphthalenone, 7-methoxy-2,3,4-trihydronaphthalen-1-one, 7-methoxytetralone, 7-methoxy-tetralone
COC1=CC2=C(CCCC2=O)C=C1
176.21
176.21
97%
Safety and Handling

Safety and Handling

Product Identifier
  • 7-Methoxy-1-tetralone

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS