missing translation for 'onlineSavingsMsg'
Learn More

7-iodo-2,3-dihydrobenzo[b]furan, 97%, Thermo Scientific™

Catalog Number p-3881438
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
264617-03-0
C8H7IO
246.047
MFCD11109311
IASDAJCCAFGOCE-UHFFFAOYSA-N
7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo
33589431
7-iodo-2,3-dihydro-1-benzofuran
C1COC2=C1C=CC=C2I
This item is not returnable. View return policy
264617-03-0
C8H7IO
246.047
MFCD11109311
IASDAJCCAFGOCE-UHFFFAOYSA-N
7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo
33589431
7-iodo-2,3-dihydro-1-benzofuran
C1COC2=C1C=CC=C2I
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 246.04
Percent Purity 97%
Chemical Name or Material 7-Iodo-2, 3-dihydrobenzo[b]furan
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'