missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyisoquinoline, 97%, Thermo Scientific Chemicals

Catalog Number p-7058471
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
7651-83-4
C9H7NO
145.16
MFCD00456131
WCRKBMABEPCYII-UHFFFAOYSA-N
7-hydroxyisoquinoline, 7-isoquinolinol, 7-hydroxy-isoquinoline, isoquinolin-7-ol, 7-hydroxy-2-azanaphthalene, pubchem6264, acmc-1bfxl, ksc380o3r, 7-hydroxyisoquinoline 250mg
459767
isoquinolin-7-ol
OC1=CC=C2C=CN=CC2=C1
This item is not returnable. View return policy
7651-83-4
C9H7NO
145.16
MFCD00456131
WCRKBMABEPCYII-UHFFFAOYSA-N
7-hydroxyisoquinoline, 7-isoquinolinol, 7-hydroxy-isoquinoline, isoquinolin-7-ol, 7-hydroxy-2-azanaphthalene, pubchem6264, acmc-1bfxl, ksc380o3r, 7-hydroxyisoquinoline 250mg
459767
isoquinolin-7-ol
OC1=CC=C2C=CN=CC2=C1

7-Hydroxyisoquinoline is used to study its photophysial behavior in polymer matrices such as polymethyl methacrylate, cellulose acetate, polyvinyl alcohol and sulfonated tetrafluoroethylene based fluoropolymer- copolymer. It also used to prepare quinine. Further, it is used as the material for all optical switching due to its excited state proton transfer (ESPT) effect.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Hydroxyisoquinoline is used to study its photophysial behavior in polymer matrices such as polymethyl methacrylate, cellulose acetate, polyvinyl alcohol and sulfonated tetrafluoroethylene based fluoropolymer- copolymer. It also used to prepare quinine. Further, it is used as the material for all optical switching due to its excited state proton transfer (ESPT) effect.

Solubility
Soluble in methanol and concentrated sulfuric acid.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 226°C to 228°C
Quantity 250 mg
Beilstein 113224
Solubility Information Soluble in methanol and concentrated sulfuric acid.
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material 7-Hydroxyisoquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'