missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxycoumarinyl-4-acetic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-3771105
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6950-82-9
C11H8O5
220.18
MFCD00037563
BNHPMQBVNXMPDU-UHFFFAOYSA-N
7-hydroxycoumarin-4-acetic acid, 2-7-hydroxy-2-oxo-2h-chromen-4-yl acetic acid, umbelliferone-4-acetic acid, 7-hydroxy-2-oxo-2h-1-benzopyran-4-acetic acid, chembl12253, 2-7-hydroxy-2-oxochromen-4-yl acetic acid, 7-hydroxy-4-coumarinylacetic acid, 2h-1-benzopyran-4-acetic acid, 7-hydroxy-2-oxo, 7-hydroxycoumarin-4-yl acetic acid, 7-hydroxy-2-oxochromen-4-yl acetic acid
5338490
2-(7-hydroxy-2-oxochromen-4-yl)acetic acid
C1=CC2=C(C=C1O)OC(=O)C=C2CC(=O)O
This item is not returnable. View return policy
6950-82-9
C11H8O5
220.18
MFCD00037563
BNHPMQBVNXMPDU-UHFFFAOYSA-N
7-hydroxycoumarin-4-acetic acid, 2-7-hydroxy-2-oxo-2h-chromen-4-yl acetic acid, umbelliferone-4-acetic acid, 7-hydroxy-2-oxo-2h-1-benzopyran-4-acetic acid, chembl12253, 2-7-hydroxy-2-oxochromen-4-yl acetic acid, 7-hydroxy-4-coumarinylacetic acid, 2h-1-benzopyran-4-acetic acid, 7-hydroxy-2-oxo, 7-hydroxycoumarin-4-yl acetic acid, 7-hydroxy-2-oxochromen-4-yl acetic acid
5338490
2-(7-hydroxy-2-oxochromen-4-yl)acetic acid
C1=CC2=C(C=C1O)OC(=O)C=C2CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 209°C to 210°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 220.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxycoumarinyl-4-acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 7-Hydroxycoumarinyl-4-acetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'