missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxycoumarin, 98%, Thermo Scientific Chemicals

Catalog Number p-7058285
24.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-35-6
C9H6O3
162.14
MFCD00006878
ORHBXUUXSCNDEV-UHFFFAOYSA-N
7-hydroxycoumarin, umbelliferone, 7-hydroxy-2h-chromen-2-one, hydrangin, skimmetin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangine, skimmetine, umbelliferon
5281426
CHEBI:27510
7-hydroxychromen-2-one
OC1=CC=C2C=CC(=O)OC2=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-35-6
C9H6O3
162.14
MFCD00006878
ORHBXUUXSCNDEV-UHFFFAOYSA-N
7-hydroxycoumarin, umbelliferone, 7-hydroxy-2h-chromen-2-one, hydrangin, skimmetin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangine, skimmetine, umbelliferon
5281426
CHEBI:27510
7-hydroxychromen-2-one
OC1=CC=C2C=CC(=O)OC2=C1

7-Hydroxycoumarin is used as an intracellular pH sensitive fluorescent indicator & bloodbrain barrier probe. It is used as a sunscreen agent, and an optical brightener for textiles. It has also been used as a gain medium for dye lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Hydroxycoumarin is used as an intracellular pH sensitive fluorescent indicator & bloodbrain barrier probe. It is used as a sunscreen agent, and an optical brightener for textiles. It has also been used as a gain medium for dye lasers.

Solubility
Soluble in water (slightly at 20°C), 95% ethanol (50 mg/ml), chloroform, acetone, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 228°C to 232°C
Quantity 25 g
Beilstein 127683
Merck Index 14,9847
Solubility Information Soluble in water (slightly at 20°C),95% ethanol (50mg/ml),chloroform,acetone,and methanol.
Formula Weight 162.14
Percent Purity 98%
Chemical Name or Material 7-Hydroxycoumarin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'