missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-PIPAT maleate, Tocris Bioscience™

Brand:  Tocris (Bio-Techne) 071910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13437893

  • £174.93 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This ligand has uniquely high affinity and selectivity for the D3 receptor. 8-Hydroxy-PIPAT maleate (Cat. No. 0797) also available.
Specifications

Chemical Identifiers

200722-46-9
487.334
7-hydroxy-pipat maleate, unii-977jc7ev31, 7-hydroxy-pipat maleate, +/-, 2-2e-3-iodoallyl propyl amino tetralin-7-ol maleate, 2-2e-3-iodoprop-2-en-1-yl propyl amino tetralin-7-ol maleate, 7-hydroxy-2-n-n-propyl-n-3'-iodo-2'-propenyl amino tetralin maleate, 7-2e-3-iodoprop-2-en-1-yl propyl amino-5,6,7,8-tetrahydronaphthalen-2-ol maleate, 2-naphthalenol, 5,6,7,8-tetrahydro-7-2e-3-iodo-2-propenyl propylamino-, 2z-2-butenedioate 1:1 salt, 7-2e-3-iodoprop-2-en-1-yl propyl amino-5,6,7,8-tetrahydronaphthalen-2-ol 2z-but-2-enedioate, 7-2e-3-iodoprop-2-en-1-yl propyl amino-5,6,7,8-tetrahydronaphthalen-2-ol 2z-but-2-enedioate salt
CHEBI:64134
CCCN(CC=CI)C1CCC2=C(C1)C=C(C=C2)O.C(=CC(=O)O)C(=O)O
C20H26INO5
IQRDHLSQXOATHD-HBRCEPSSSA-N
24978534
(Z)-but-2-enedioic acid;7-[[(E)-3-iodoprop-2-enyl]-propylamino]-5,6,7,8-tetrahydronaphthalen-2-ol

Specifications

10mg
200722-46-9
7-hydroxy-pipat maleate, unii-977jc7ev31, 7-hydroxy-pipat maleate, +/-, 2-2e-3-iodoallyl propyl amino tetralin-7-ol maleate, 2-2e-3-iodoprop-2-en-1-yl propyl amino tetralin-7-ol maleate, 7-hydroxy-2-n-n-propyl-n-3'-iodo-2'-propenyl amino tetralin maleate, 7-2e-3-iodoprop-2-en-1-yl propyl amino-5,6,7,8-tetrahydronaphthalen-2-ol maleate, 2-naphthalenol, 5,6,7,8-tetrahydro-7-2e-3-iodo-2-propenyl propylamino-, 2z-2-butenedioate 1:1 salt, 7-2e-3-iodoprop-2-en-1-yl propyl amino-5,6,7,8-tetrahydronaphthalen-2-ol 2z-but-2-enedioate, 7-2e-3-iodoprop-2-en-1-yl propyl amino-5,6,7,8-tetrahydronaphthalen-2-ol 2z-but-2-enedioate salt
CCCN(CC=CI)C1CCC2=C(C1)C=C(C=C2)O.C(=CC(=O)O)C(=O)O
487.334
CHEBI:64134
>98%
7-Hydroxy-PIPAT maleate
C20H26INO5
IQRDHLSQXOATHD-HBRCEPSSSA-N
(Z)-but-2-enedioic acid;7-[[(E)-3-iodoprop-2-enyl]-propylamino]-5,6,7,8-tetrahydronaphthalen-2-ol
24978534
487.33
Videos
Documents