Learn More
7-Hydroxy-DPAT hydrobromide, Tocris Bioscience™
Dopamine agonist (D3 ≥ D2 < > D4)
Brand:  Tocris (Bio-Techne) 0706/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13457883
  • £83.79 / 10mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
D3 dopamine receptor agonist (Ki values are ∽ 1, 10, 650 and ∽ 5000 nM for D3, D2, D4 and D1 receptors respectively).
8-Hydroxy Isomer also available.
Specifications
Chemical Identifiers
76135-30-3
328.294
(±)-7-Hydroxy-2-dipropylaminotetralin hydrobromide
7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
C16H26BrNO
ODNDMTWHRYECKX-UHFFFAOYSA-N
11957566
CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br
Specifications
10mg
76135-30-3
(±)-7-Hydroxy-2-dipropylaminotetralin hydrobromide
CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br
328.294
328.28
7-Hydroxy-DPAT hydrobromide
C16H26BrNO
ODNDMTWHRYECKX-UHFFFAOYSA-N
7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
11957566
>99%