missing translation for 'onlineSavingsMsg'
Learn More

7-hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid, Maybridge

Brand:  Maybridge BTB10150DA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 105212-09-7 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10091782

  • £50.00 / 1g


Estimated Shipment date
from Supplier 27-10-2017
Add to basket

Specifications

Chemical Identifiers

7-Hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid
97%
248.23
97%
C13H12O5

Specifications

7-Hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid
105212-09-7
97%
248.23
1g
97%
C13H12O5
Documentation

Technical Documents