missing translation for 'onlineSavingsMsg'
Learn More

7-Deazaguanine, 90+%, Thermo Scientific™

Catalog Number p-4813286
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7355-55-7
C6H6N4O
150.14
MFCD09033259
OLAFFPNXVJANFR-UHFFFAOYSA-N
7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
96253
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
NC1=NC(=O)C2=C(NC=C2)N1
This item is not returnable. View return policy
7355-55-7
C6H6N4O
150.14
MFCD09033259
OLAFFPNXVJANFR-UHFFFAOYSA-N
7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
96253
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
NC1=NC(=O)C2=C(NC=C2)N1
Melting Point >230.0°C
Assay Percent Range 90% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 150.14
Percent Purity 90+%
Chemical Name or Material 7-Deazaguanine
compliance-icons
Product Identifier
  • 7-Deazaguanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'