Learn More
7-Chlorokynurenic acid, Tocris Bioscience™
Potent competitive inhibitor of L-glutamate uptake
Brand:  Tocris (Bio-Techne) 0237/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13437503
  • £316.05 / 50mg
Estimated Shipment date
from Supplier 23-10-2019
Description
Description
NMDA receptor antagonist acting at the glycine site. Potent competitive inhibitor of L-glutamate transport into synaptic vesicles.
Available as part of the NMDA Receptor - Glycine Site Tocriset™ . Sodium Salt also available.
Specifications
Chemical Identifiers
18000-24-3
223.612
7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
CHEBI:63965
C1=CC2=C(C=C1Cl)NC(=CC2=O)C(=O)O
C10H6ClNO3
UAWVRVFHMOSAPU-UHFFFAOYSA-N
1884
7-chloro-4-oxo-1H-quinoline-2-carboxylic acid
Specifications
50mg
18000-24-3
7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
C1=CC2=C(C=C1Cl)NC(=CC2=O)C(=O)O
223.612
CHEBI:63965
>98%
7-Chlorokynurenic acid
C10H6ClNO3
UAWVRVFHMOSAPU-UHFFFAOYSA-N
7-chloro-4-oxo-1H-quinoline-2-carboxylic acid
1884
223.62