missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-4-hydroxyquinoline, 99%, Thermo Scientific™

Catalog Number p-4813282
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
86-99-7
C9H6ClNO
179.61
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol, 7-chloro-4-hydroxyquinoline, 7-chloro-4-quinolinol, 7-chloroquinolin-4 1h-one, 4-quinolinol, 7-chloro, 4 1h-quinolinone, 7-chloro, 7-chloro-quinolin-4-ol, 7-chloro-4 1h-quinolinone, 4mrw, 7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O
This item is not returnable. View return policy
86-99-7
C9H6ClNO
179.61
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol, 7-chloro-4-hydroxyquinoline, 7-chloro-4-quinolinol, 7-chloroquinolin-4 1h-one, 4-quinolinol, 7-chloro, 4 1h-quinolinone, 7-chloro, 7-chloro-quinolin-4-ol, 7-chloro-4 1h-quinolinone, 4mrw, 7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 276.0°C to 279.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 179.61
Percent Purity 99%
Chemical Name or Material 7-Chloro-4-hydroxyquinoline
compliance-icons
Product Identifier
  • 7-Chloro-4-hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'