missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 7-Chloro-3-(2-hydroxyethyl)-1H-pyrazolo[4,3-d]pyrimidine, 97%

Brand:  Alfa Aesar™ H63802.MD

Code : NEW

Additional Details : CAS Number : 64127-15-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15498526

  • £24.70 / 250mg


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

7-Chloro-3-(2-hydroxyethyl)-1H-pyrazolo[4,3-d]pyrimidine
198.61
2-(7-Chloro-3H-pyrazolo[4,3-d]pyrimidin-3-yl)ethanol
64127-15-7
MFCD12022632

Specifications

7-Chloro-3-(2-hydroxyethyl)-1H-pyrazolo[4,3-d]pyrimidine
64127-15-7
97%
MFCD12022632
250mg
Solid
198.61
2-(7-Chloro-3H-pyrazolo[4,3-d]pyrimidin-3-yl)ethanol
Videos
SDS