missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Quantity | 1 g |
Formula Weight | 204.1 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'