missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydro-1,5-benzodioxepin, 96%, Thermo Scientific Chemicals

Catalog Number p-7030061
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
147644-11-9
C9H9BrO2
229.073
MFCD00508291
AZCHNKNSOZJHSH-UHFFFAOYSA-N
2776394
7-bromo-3,4-dihydro-2H-1,5-benzodioxepine
C1COC2=C(C=C(C=C2)Br)OC1
This item is not returnable. View return policy
147644-11-9
C9H9BrO2
229.073
MFCD00508291
AZCHNKNSOZJHSH-UHFFFAOYSA-N
2776394
7-bromo-3,4-dihydro-2H-1,5-benzodioxepine
C1COC2=C(C=C(C=C2)Br)OC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 145°C to 148°C (16 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.579
Quantity 1 g
Synonym 7-bromo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin, 7-bromo-3,4-dihydro, acmc-1c5pl, 7-bromo-2h,3h,4h-benzo b 1,4-dioxepin, 7-bromo-3,4-dihydro-2h-1,5-benzodioxepin, 2h-1,5-benzodioxepin,7-bromo-3,4-dihydro, 7-bromo-3,4-dihydro-1,5-benzodioxepin
Formula Weight 229.07
Percent Purity 96%
Chemical Name or Material 7-Bromo-3,4-dihydro-1,5-benzodioxepin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'