missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Bromo-3,4-dihydro-1(2H)-naphthalenone, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 225.08 |
Percent Purity | 97% |
Chemical Name or Material | 7-Bromo-3, 4-dihydro-1(2H)-naphthalenone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'