missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-methyl-1H-indole, 97%, Thermo Scientific™

Catalog Number p-3586830
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
280752-68-3
C9H8BrN
210.07
MFCD09879960
CALOMHQMSHLUJX-UHFFFAOYSA-N
7-bromo-1-methyl-1h-indole, 1h-indole,7-bromo-1-methyl, 1h-indole, 7-bromo-1-methyl, 1-methyl-7-bromoindole, 1-methyl-7-bromo-1h-indole
21950069
7-bromo-1-methylindole
CN1C=CC2=CC=CC(Br)=C12
This item is not returnable. View return policy
280752-68-3
C9H8BrN
210.07
MFCD09879960
CALOMHQMSHLUJX-UHFFFAOYSA-N
7-bromo-1-methyl-1h-indole, 1h-indole,7-bromo-1-methyl, 1h-indole, 7-bromo-1-methyl, 1-methyl-7-bromoindole, 1-methyl-7-bromo-1h-indole
21950069
7-bromo-1-methylindole
CN1C=CC2=CC=CC(Br)=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.07
Percent Purity 97%
Chemical Name or Material 7-Bromo-1-methyl-1H-indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'