missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole, 98%, Thermo Scientific Chemicals

Catalog Number 10539620
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
271-63-6
C7H6N2
118.14
MFCD00005606,MFCD00075874
MVXVYAKCVDQRLW-UHFFFAOYSA-N
7-azaindole, 1h-pyrrolo 2,3-b pyridine, 1,7-diazaindene, 7h-pyrrolo 2,3-b pyridine, 7-aza-1-pyrindine, 1,7-dideazapurine, 1 h-pyrrolo 2,3:b pyridine, 7-aza indole, unii-qx4465nr9t
9222
1H-pyrrolo[2,3-b]pyridine
N1C=CC2=CC=CN=C12
This item is not returnable. View return policy
271-63-6
C7H6N2
118.14
MFCD00005606,MFCD00075874
MVXVYAKCVDQRLW-UHFFFAOYSA-N
7-azaindole, 1h-pyrrolo 2,3-b pyridine, 1,7-diazaindene, 7h-pyrrolo 2,3-b pyridine, 7-aza-1-pyrindine, 1,7-dideazapurine, 1 h-pyrrolo 2,3:b pyridine, 7-aza indole, unii-qx4465nr9t
9222
1H-pyrrolo[2,3-b]pyridine
N1C=CC2=CC=CN=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 103.0°C to 107.0°C
Color White
Boiling Point 270°C (1004.0 HPA)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, V,6, 306
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetonitrile
Formula Weight 118.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 7-Azaindole
Show More Show Less
compliance-icons
Product Identifier
  • 7-Azaindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'