missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 15405947
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
479553-01-0
C8H6N2O2
162.148
MFCD08272236
HPPPHDPVSYYWCH-UHFFFAOYSA-N
1h-pyrrolo 2,3-b pyridine-4-carboxylic acid, 7-azaindole-4-carboxylic acid, 7-azaindole-4-carboxylic acid., 1h-pyrrolo 2,3-b pyridine-4 carboxylic acid
22477426
1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid
C1=CNC2=NC=CC(=C21)C(=O)O
This item is not returnable. View return policy
479553-01-0
C8H6N2O2
162.148
MFCD08272236
HPPPHDPVSYYWCH-UHFFFAOYSA-N
1h-pyrrolo 2,3-b pyridine-4-carboxylic acid, 7-azaindole-4-carboxylic acid, 7-azaindole-4-carboxylic acid., 1h-pyrrolo 2,3-b pyridine-4 carboxylic acid
22477426
1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid
C1=CNC2=NC=CC(=C21)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >330°C
Color White to Cream
Quantity 250 mg
Formula Weight 162.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Azaindole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'