missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dihydroxyflavone, Tocris Bioscience™

TrkB agonist

Brand:  Tocris (Bio-Techne) 382610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13433664

  • £149.94 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Tyrosine kinase receptor B (TrkB) agonist that binds to the extracellular domain of the receptor (Kd = 320 nM). Inhibits glutamate-triggered apoptosis in hippocampal neurons in vitro and in vivo. Exhibits neuroprotective effects in an HD mouse model.
Specifications

Chemical Identifiers

38183-03-8
254.241
7,8-dihydroxyflavone, 7,8-dihydroxy-2-phenyl-4h-chromen-4-one, 7,8-dihydroxy-flavone, 4h-1-benzopyran-4-one, 7,8-dihydroxy-2-phenyl, 7,8-dhf, 7,8-dihydroxy-2-phenyl-4-benzopyrone, 7,8-dihydroxy-2-phenyl-chromen-4-one, 7,8-dihydroxyflavone hydrate, chembl75267, 7,8-dihydroxy-2-phenyl-4h-1-benzopyran-4-one
7,8-dihydroxy-2-phenylchromen-4-one
C15H10O4
COCYGNDCWFKTMF-UHFFFAOYSA-N
1880
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)O)O

Specifications

10mg
38183-03-8
7,8-dihydroxyflavone, 7,8-dihydroxy-2-phenyl-4h-chromen-4-one, 7,8-dihydroxy-flavone, 4h-1-benzopyran-4-one, 7,8-dihydroxy-2-phenyl, 7,8-dhf, 7,8-dihydroxy-2-phenyl-4-benzopyrone, 7,8-dihydroxy-2-phenyl-chromen-4-one, 7,8-dihydroxyflavone hydrate, chembl75267, 7,8-dihydroxy-2-phenyl-4h-1-benzopyran-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)O)O
254.241
254.24
7, 8-Dihydroxyflavone
C15H10O4
COCYGNDCWFKTMF-UHFFFAOYSA-N
7,8-dihydroxy-2-phenylchromen-4-one
1880
>98%
Videos
Documents