missing translation for 'onlineSavingsMsg'
Learn More

7,7,8,8-Tetracyanoquinodimethane, 98%, Thermo Scientific Chemicals

Catalog Number 10668894
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1518-16-7
C12H4N4
204.19
MFCD00011664
PCCVSPMFGIFTHU-UHFFFAOYSA-N
7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
73697
CHEBI:52445
2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N
This item is not returnable. View return policy
1518-16-7
C12H4N4
204.19
MFCD00011664
PCCVSPMFGIFTHU-UHFFFAOYSA-N
7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
73697
CHEBI:52445
2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 287°C to 289°C
Color Brown-Orange to Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Fieser 01,1136; 12,464
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetomitrile,tetrahydrofuran and,dioxane
Formula Weight 204.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 7,7,8,8-Tetracyanoquinodimethane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 7,7,8,8-Tetracyanoquinodimethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'