missing translation for 'onlineSavingsMsg'
Learn More

7,7,8,8-Tetracyanoquinodimethane, 98%, ACROS Organics™

Brand:  Acros Organics 169360100

Code : 29

Additional Details : CAS Number : 1518-16-7 Weight : 0.06000kg Transport : UN number : 3439 Chem class : 1 Pack group : III


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10636762

  • £121.20 / 10g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

7,7,8,8-Tetracyanoquinodimethane
1518-16-7
100.0
98%
204.19
2,5-Cyclohexadiene-δ1,α.4,α'-dimalononitrile, TCNQ
98%
97.5
≥97.5%
C12H4N4
MFCD00011664

Specifications

7,7,8,8-Tetracyanoquinodimethane
10g
1518-16-7
100.0
Glass bottle
≥97.5%
C12H4N4
MFCD00011664
2,5-Cyclohexadiene-δ1,α.4,α'-dimalononitrile, TCNQ
Orange-brown to green
98%
97.5
Authentic
Crystalline Powder
98%
204.19
01,1136; 12,464
Videos
SDS