missing translation for 'onlineSavingsMsg'
Learn More

7,12-Dimethylbenz[a]anthracene, 98%, Thermo Scientific Chemicals

Catalog Number p-116440
25.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 mg
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
57-97-6
C20H16
256.34
MFCD00003600
ARSRBNBHOADGJU-UHFFFAOYSA-N
dmba, 7,12-dimethylbenz a anthracene, 7,12-dimethylbenzanthracene, 9,10-dimethyl-1,2-benzanthracene, 7,12-dmba, 7,12-dimethylbenzo a anthracene, 7,12-dimethylbenzanthrancene, 6,7-dimethyl-1,2-benzanthracene, 1,4-dimethyl-2,3-benzphenanthrene
6001
CHEBI:254496
7,12-dimethylbenzo[a]anthracene
CC1=C2C=CC3=CC=CC=C3C2=C(C4=CC=CC=C14)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
57-97-6
C20H16
256.34
MFCD00003600
ARSRBNBHOADGJU-UHFFFAOYSA-N
dmba, 7,12-dimethylbenz a anthracene, 7,12-dimethylbenzanthracene, 9,10-dimethyl-1,2-benzanthracene, 7,12-dmba, 7,12-dimethylbenzo a anthracene, 7,12-dimethylbenzanthrancene, 6,7-dimethyl-1,2-benzanthracene, 1,4-dimethyl-2,3-benzphenanthrene
6001
CHEBI:254496
7,12-dimethylbenzo[a]anthracene
CC1=C2C=CC3=CC=CC=C3C2=C(C4=CC=CC=C14)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 122°C to 125°C
Color Green to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 05, III, 2413
Merck Index 15, 3257
Formula Weight 256.34
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 7, 12-Dimethylbenz[a]anthracene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 7,12-Dimethylbenz¢a!anthracene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'