missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%

Brand:  Alfa Aesar™ H63138.03

Code : NEW

Additional Details : CAS Number : 131747-42-7 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15409668

  • £58.00 / 1g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

6-(Trifluoromethyl)pyridine-2-carboxylic acid
191.11
UN2811
131747-42-7
MFCD02180819
6-(Trifluoromethyl)picolinic acid

Specifications

6-(Trifluoromethyl)pyridine-2-carboxylic acid
154° to 158°C
131747-42-7
98%
MFCD02180819
6-(Trifluoromethyl)picolinic acid
Off-white
1g
Solid
191.11
UN2811
Videos
SDS