missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Thioguanine, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Description
6-Thioguanine acts as an antineoplastic and purine antimetabolite. It is also useful as an inhibition of stimulated expression of TNF-related apoptosis-inducing ligand (TRAIL) protein. It is involved in the treatment of acute leukemias and Psoriasis. Further, it is used for the treatment of ulcerative colitis and autoimmune diseases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | >360°C |
Assay Percent Range | 0.98 |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 157765 |
Merck Index | 14,9337 |
Solubility Information | Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water. |
IUPAC Name | 2-amino-3,7-dihydropurine-6-thione |
Formula Weight | 167.19 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'