missing translation for 'onlineSavingsMsg'
Learn More

6-(Tetrahydropyran-4-yloxy)nicotinonitrile, 97%, Thermo Scientific™

Catalog Number p-3830134
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
884507-60-2
C11H12N2O2
204.23
MFCD09702388
CCEWHGWDICBASG-UHFFFAOYSA-N
6-tetrahydropyran-4-yloxy nicotinonitrile, 6-tetrahydro-2h-pyran-4-yloxy pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yloxy pyridine-3-carbonitrile, 6-oxan-4-yl oxy pyridine-3-carbonitrile, 6-tetrahydro-2h-pyran-4-yl oxy nicotinonitrile, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-3-carbonitrile
24229580
6-(oxan-4-yloxy)pyridine-3-carbonitrile
N#CC1=CN=C(OC2CCOCC2)C=C1
This item is not returnable. View return policy
884507-60-2
C11H12N2O2
204.23
MFCD09702388
CCEWHGWDICBASG-UHFFFAOYSA-N
6-tetrahydropyran-4-yloxy nicotinonitrile, 6-tetrahydro-2h-pyran-4-yloxy pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yloxy pyridine-3-carbonitrile, 6-oxan-4-yl oxy pyridine-3-carbonitrile, 6-tetrahydro-2h-pyran-4-yl oxy nicotinonitrile, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-3-carbonitrile
24229580
6-(oxan-4-yloxy)pyridine-3-carbonitrile
N#CC1=CN=C(OC2CCOCC2)C=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.23
Percent Purity 97%
Chemical Name or Material 6-(Tetrahydropyran-4-yloxy)nicotinonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'