missing translation for 'onlineSavingsMsg'
Learn More

6-Quinoxalinecarboxylic acid, 95%, Maybridge

Brand:  Maybridge CC11701EE

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 6925-00-4 Weight : 0.10000kg

Quantity 50g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10707095

  • £376.00 / 50g


Estimated Shipment date
from Supplier 28-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

6-Quinoxalinecarboxylic acid
95%
174.16
95%
C9H6N2O2

Specifications

6-Quinoxalinecarboxylic acid
Amber Glass Bottle
95%
174.16
6925-00-4
95%
C9H6N2O2
50g
Documentation

Technical Documents