missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinecarboxylic Acid, 98%, Thermo Scientific Chemicals

Catalog Number 10454615
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
10349-57-2
C10H7NO2
173.17
MFCD00047613
VXGYRCVTBHVXMZ-UHFFFAOYSA-N
6-quinolinecarboxylic acid, pubchem7578, quinoline-6-carboylic acid, quinolin-6-carboxylic acid, 6-quinoline carboxylic acid, 6-quinoline-carboxylic acid, ksc177e5h, vxgyrcvtbhvxmz-uhfffaoysa, 2-3-cyclopentyl-2-oxo-2,3-dihydro-1h-imidazo 4,5-b pyridin-1-yl-n-2,3-dihydro-1,4-benzodioxin-6-yl acetamide
82571
quinoline-6-carboxylic acid
C1=CC2=C(C=CC(=C2)C(=O)O)N=C1
This item is not returnable. View return policy
10349-57-2
C10H7NO2
173.17
MFCD00047613
VXGYRCVTBHVXMZ-UHFFFAOYSA-N
6-quinolinecarboxylic acid, pubchem7578, quinoline-6-carboylic acid, quinolin-6-carboxylic acid, 6-quinoline carboxylic acid, 6-quinoline-carboxylic acid, ksc177e5h, vxgyrcvtbhvxmz-uhfffaoysa, 2-3-cyclopentyl-2-oxo-2,3-dihydro-1h-imidazo 4,5-b pyridin-1-yl-n-2,3-dihydro-1,4-benzodioxin-6-yl acetamide
82571
quinoline-6-carboxylic acid
C1=CC2=C(C=CC(=C2)C(=O)O)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 293.0°C to 294.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 100 g
Beilstein 22, V,3, 214
Formula Weight 173.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Quinolinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Quinolinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'