missing translation for 'onlineSavingsMsg'
Learn More

6-Pyrrolidin-1-ylpyridine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3522561
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
450368-20-4
C10H12N2O2
192.22
MFCD09064946
HXPHLYQBFPDHNK-UHFFFAOYSA-N
23509740
OC(=O)C1=NC(=CC=C1)N1CCCC1
This item is not returnable. View return policy
450368-20-4
C10H12N2O2
192.22
MFCD09064946
HXPHLYQBFPDHNK-UHFFFAOYSA-N
23509740
OC(=O)C1=NC(=CC=C1)N1CCCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 6-1-pyrrolidyl pyridine-2-carboxylic acid, 6-pyrrolidin-1-yl picolinic acid, 6-pyrrolidin-1-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-1-pyrrolidinyl, 6-pyrrolidinylpyridine-2-carboxylic acid, 6-1-pyrrolidyl pyridine-2-carboxylicacid, 2-pyridinecarboxylicacid,6-1-pyrrolidinyl, 6-1-pyrrolidinyl-2-pyridinecarboxylic acid
IUPAC Name 6-(pyrrolidin-1-yl)pyridine-2-carboxylic acid
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material 6-Pyrrolidin-1-ylpyridine-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'