missing translation for 'onlineSavingsMsg'
Learn More

(6-Pyrrolidin-1-ylpyrid-2-yl)methylamine, 97%, Maybridge

Brand:  Maybridge CC38013CB

Code : 31

Additional Details : CAS Number : 868755-49-1 Weight : 0.05025kg Transport : UN number : 2922 Chem class : 8 Pack group : III

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10286393

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(6-Pyrrolidin-1-ylpyrid-2-yl)methylamine
96.5
95%
C10H15N3
MFCD09064947
868755-49-1
100.0
95%
177.25

Specifications

(6-Pyrrolidin-1-ylpyrid-2-yl)methylamine
868755-49-1
100.0
95%
C10H15N3
MFCD09064947
250mg
96.5
Amber Glass Bottle
95%
177.25
Videos
SDS