missing translation for 'onlineSavingsMsg'
Learn More

(6-Piperidinopyrid-2-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-3484508
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
869901-07-5
C11H16N2O
192.262
MFCD08690242
WLHPFWHASVOLSO-UHFFFAOYSA-N
6-piperidinopyrid-2-yl methanol, 6-piperidin-1-yl pyridin-2-yl methanol, 6-piperidyl-2-pyridyl methan-1-ol, 2-pyridinemethanol,6-1-piperidinyl, 6-piperidin-1-ylpyridin-2-yl methanol, 2-hydroxymethyl-6-piperidin-1-ylpyridine
18525733
(6-piperidin-1-ylpyridin-2-yl)methanol
C1CCN(CC1)C2=CC=CC(=N2)CO
This item is not returnable. View return policy
869901-07-5
C11H16N2O
192.262
MFCD08690242
WLHPFWHASVOLSO-UHFFFAOYSA-N
6-piperidinopyrid-2-yl methanol, 6-piperidin-1-yl pyridin-2-yl methanol, 6-piperidyl-2-pyridyl methan-1-ol, 2-pyridinemethanol,6-1-piperidinyl, 6-piperidin-1-ylpyridin-2-yl methanol, 2-hydroxymethyl-6-piperidin-1-ylpyridine
18525733
(6-piperidin-1-ylpyridin-2-yl)methanol
C1CCN(CC1)C2=CC=CC(=N2)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material (6-Piperidinopyrid-2-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'