missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylpyrid-3-ylamine, 97%, Maybridge

Brand:  Maybridge CC21114DA

Code : 31

Additional Details : CAS Number : 126370-67-0 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10537854

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

6-Phenylpyrid-3-ylamine
96.5
97%
C11H10N2
MFCD04114197
126370-67-0
100.0
97%
170.21

Specifications

6-Phenylpyrid-3-ylamine
126370-67-0
Amber Glass Bottle
97%
C11H10N2
MFCD04114197
1g
96.5
100.0
97%
170.21
Videos
SDS