missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylpyrid-3-ylamine, 97%, Thermo Scientific™

Catalog Number p-3522559
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
126370-67-0
C11H10N2
170.22
MFCD04114197
FLIQYTXJLWGVBG-UHFFFAOYSA-N
6-phenyl-3-pyridinamine, 5-amino-2-phenylpyridine, 6-phenylpyrid-3-ylamine, 3-amino-6-phenylpyridine, 3-pyridinamine,6-phenyl, 6-phenyl-pyridin-3-ylamine, 6-phenylpyridine-3-amine, 6-phenyl-3-pyridylamine, acmc-1c4ss, 5-amino-2-phenyl pyridine
2762877
6-phenylpyridin-3-amine
NC1=CN=C(C=C1)C1=CC=CC=C1
This item is not returnable. View return policy
126370-67-0
C11H10N2
170.22
MFCD04114197
FLIQYTXJLWGVBG-UHFFFAOYSA-N
6-phenyl-3-pyridinamine, 5-amino-2-phenylpyridine, 6-phenylpyrid-3-ylamine, 3-amino-6-phenylpyridine, 3-pyridinamine,6-phenyl, 6-phenyl-pyridin-3-ylamine, 6-phenylpyridine-3-amine, 6-phenyl-3-pyridylamine, acmc-1c4ss, 5-amino-2-phenyl pyridine
2762877
6-phenylpyridin-3-amine
NC1=CN=C(C=C1)C1=CC=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material 6-Phenylpyrid-3-ylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'