missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenylnicotinonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 6-phenylnicotinonitrile, 5-cyano-2-phenylpyridine, 3-pyridinecarbonitrile,6-phenyl, 3-pyridinecarbonitrile, 6-phenyl, 6-phenylniocotinonitrile, 2-phenyl-5-cyanopyridine, 3-cyano-6-phenylpyridine, 6-phenyl-3-pyridinecarbonitrile |
Formula Weight | 180.21 |
Percent Purity | 97% |
Chemical Name or Material | 6-Phenylnicotinonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'